Learn More
MSC Thomas & Betts TV10-10F-XV #10 Stud, 12 to 10 AWG Compatible, Partially Insulated, Crimp Connection, Standard Fork Terminal 0.283" Fork Width, Vinyl Insulation, Yellow, 1.004" Overall Length, 600 Volts, 0.039" Thick, Copper Contacts
Catalog No. 501296009
Encompass
Thomas & Betts TV10-10F-XV #10 Stud, 12 to 10 AWG Compatible, Partially Insulated, Crimp Connection, Standard Fork Terminal 0.283" Fork Width, Vinyl Insulation, Yellow, 1.004" Overall Length, 600 Volts, 0.039" Thick, Copper Contacts
Catalog No. 50-129-6009 Supplier MSC Supplier No. 68886829
Thomas & Betts TV10-10F-XV #10 Stud, 12 to 10 AWG Compatible, Partially Insulated, Crimp Connection, Standard Fork Terminal 0.283" Fork Width, Vinyl Insulation, Yellow, 1.004" Overall Length, 600 Volts, 0.039" Thick, Copper Contacts