Learn More
Sigma Aldrich 12 12 13 13 14 14 15 15 16 16 17 17 17-tridecafluoroheptadecyltrimethoxysilane
Catalog No. 503126105 Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-6105 250 mg
50-312-5885 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-312-6105 Supplier Sigma Aldrich Supplier No. SIK411620250MG
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
These products are used to form water repellent surface coating and/or lubricated surfaces on silicon, silicon oxide, etc. The fluorinated carbon gives the omniphobic properties to the surface. The coated surface is more water repellent when a product having more fluorinated carbons is used. The silane group provide a linker to covalently bond to silicon and silicon oxide surfaces. The methoxy groups are more reactive than the ethoxy ones, but they release methanol instead of ethanol for ethoxy groups.Product of SiKEMIA
Specifications
Quantity 250 mg