Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 12 12 13 13 14 14 15 15 16 16 17 17 18 18 19 19 19-heptadecafluorononadecylphosphonic Acid

Catalog No. 503125636 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-312-5636 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-312-5636 Supplier Sigma Aldrich Supplier No. SIK712210250MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

These products are used to form water repellent and/or lubricated surfaces on metal oxide materials (TiO2, ZnO, etc.).The fluorinated carbon gives the omniphobic properties to the surface. The coated surface is more water repellent when a product having more fluorinated carbons is used. The phosphonic acids groups can covalently bond to metal oxide surfaces to form sufficient self-assembled monolayer.Product of SiKEMIA

Specifications

CAS 625095-76-3
Molecular Formula C19H24F17O3P
Linear Formula C19H24F17O3P
Quantity 250 mg
Molecular Weight (g/mol) 654.34
Percent Purity 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.