Learn More
Medchemexpress LLC (±)12(13)-DiHOME-d4 | 59981-84-9 | 99.0% | C18H30D4O4 | 25 UG
SDP
Supplier:  Medchemexpress LLC 25UGHY143662S
Encompass_Preferred
(±)12(13)-DiHOME-d4 is a deuterium labeled form of (±)12(13)-DiHOME. It is used as a stable heavy isotope tracer in drug development to study pharmacokinetic and metabolic profiles, and for quantitative analysis.
  • Serves as an internal standard for quantitative analysis
  • Compatible with techniques such as NMR, GC-MS, or LC-MS
  • Incorporation of stable heavy isotopes can affect pharmacokinetic and metabolic profiles
Catalog No. 50-003-56760