Learn More
Cayman Chemical 12 13-EpOME 100UG
SDP
Catalog No. 502896337
Encompass_Preferred
The 12,13-cis epoxide form of linoleic acid; is formed primarily via linoleic acid metabolism by CYP2J2, CYP2C8, and CYP2C9, however, CYP1A1 can contribute to (±)12(13)-EpOME production when pharmacologically induced; induces mitochondrial dysfunction and cell death in renal proximal tubule epithelial cells at 500 µM
Catalog No. 50-289-6337 Supplier Cayman Chemical Supplier No. 52450100 UG
The 12,13-cis epoxide form of linoleic acid; is formed primarily via linoleic acid metabolism by CYP2J2, CYP2C8, and CYP2C9, however, CYP1A1 can contribute to (±)12(13)-EpOME production when pharmacologically induced; induces mitochondrial dysfunction and cell death in renal proximal tubule epithelial cells at 500 µM