Learn More
Medchemexpress LLC (±)12(13)Epoxy-9Z-octadecenoic-9,10,12,13-d4 acid | 99.0% | 300.47 | 50 UG
SDP
Supplier:  Medchemexpress LLC HY139959S50UG
Encompass_Preferred
(±)12(13)-EpOME-d4 is a deuterium labeled stable isotope of (±)12(13)-EpOME. It is designed for research purposes, particularly as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS.
  • Used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs
  • Appears as a colorless to light yellow liquid
  • Molecular formula: C18H28D4O3
Catalog No. 50-003-51227