Learn More
Cayman Chemical 12 13DiHOMEd4 GC MS Standards
SDP
Catalog No. 502889050
Encompass_Preferred
An internal standard for the quantification of 12(13)-DiHOME by GC- or LC-MS
Catalog No. 50-288-9050 Supplier Cayman Chemical Supplier No. 1000999425 UG
An internal standard for the quantification of 12(13)-DiHOME by GC- or LC-MS