Learn More
Cayman Chemical 12 13EpOMEd4 GC MS Standards
SDP
Catalog No. 502888647
Encompass_Preferred
An internal standard for the quantification of 12(13)-EpOME by GC- or LC-MS
Catalog No. 50-288-8647 Supplier Cayman Chemical Supplier No. 1000999650 UG
An internal standard for the quantification of 12(13)-EpOME by GC- or LC-MS