Learn More
Cayman Chemical 12 13EpOMEd4 GC MS Standards
SDP
Supplier:  Cayman Chemical 10009996100 UG
Encompass_Preferred
An internal standard for the quantification of 12(13)-EpOME by GC- or LC-MS
Catalog No. 50-288-9195