Learn More
Chemglass Life Sciences 12/2 Ball Joint, Clamp Size: 12, Tubing OD: 7.5mm
SDP
Catalog No. CG12002
Encompass_Preferred
Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps. 12/2 Ball Joint, Clamp Size: 12, Tubing OD: 7.5mm.
Catalog No. CG12002 Supplier Chemglass Life Sciences Supplier No. CG12002
Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps. 12/2 Ball Joint, Clamp Size: 12, Tubing OD: 7.5mm.