Learn More
Chemglass Life Sciences Joint, 12/2, Socket, Clamp Size 12, 7.5mm Tubing
SDP
Catalog No. CG12102
Encompass_Preferred
Joint, 12/2, Socket, Clamp Size 12, 7.5mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. CG12102 Supplier Chemglass Life Sciences Supplier No. CG12102
Joint, 12/2, Socket, Clamp Size 12, 7.5mm Tubing. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.