Learn More
Ace Glass, Inc. 12/3 BALL JOINT (GLASSBLOWER)
SDP
Catalog No. 50975265
Encompass_Preferred
Full length, standard wall 12/3 ball joint. This ACE ball joint is precision ground and vacuum leak-tested for fit and interchangeability.
Catalog No. 50-975-265 Supplier Ace Glass, Inc. Supplier No. 765504
Full length, standard wall 12/3 ball joint. This ACE ball joint is precision ground and vacuum leak-tested for fit and interchangeability.