Learn More
Chemglass Life Sciences 12/3 O-Ring Ball Joint, Clamp Size: 12, O-Ring Size: 011, Tubing OD: 8mm
SDP
Catalog No. CG12202
Encompass_Preferred
12/3 O-Ring Ball Joint, Clamp Size: 12, O-Ring Size: 011,Tubing OD: 8mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.
Catalog No. CG12202 Supplier Chemglass Life Sciences Supplier No. CG12202
12/3 O-Ring Ball Joint, Clamp Size: 12, O-Ring Size: 011,Tubing OD: 8mm. Ball joint with a Viton o-ring fitted into a groove tooled at the midpoint of the grind. Provides for a flexible, vacuum tight seal when used with CG-121-P polished socket joints. Supplied complete with one Viton o-ring of the listed size.