Learn More
JANT PHARMACAL CORPORATION Accutest ValuPak Drug Test Cup 12 with Adulterants. AMP 1000/BAR/BUP/BZO/COC 300/MDMA/MET 1000/MOP 300/MTD/OXY/PCP/THC plus Adulterant Screens (OXI/SG/PH) 25 Cups/kit box
SDP
Catalog No. NC3457088
Encompass_Preferred
Accutest CLIA Waived 12 panel drug test cup with adulterants is a rapid, one-step qualitative urine drug test that uses a visually read competitive binding immunoassay method for screening multiple drugs and drug metabolites in human urine without the need of instrumentation.
Catalog No. NC3457088 Supplier JANT PHARMACAL CORPORATION Supplier No. DS432
Accutest CLIA Waived 12 panel drug test cup with adulterants is a rapid, one-step qualitative urine drug test that uses a visually read competitive binding immunoassay method for screening multiple drugs and drug metabolites in human urine without the need of instrumentation.