Learn More
Chemglass Life Sciences 12/30 Outer Joint, Full Length, on 16mm OD x 1.2mm W tubing
SDP
Catalog No. CG10103
Encompass_Preferred
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 12/30 Outer Joint, Full Length, on 16mm OD x 1.2mm W tubing.
Catalog No. CG10103 Supplier Chemglass Life Sciences Supplier No. CG10103
Standard taper outer joint tooled on tubing approximately the same diameter as the top reinforcing ring, affording no hold-up below joint. 12/30 Outer Joint, Full Length, on 16mm OD x 1.2mm W tubing.