Learn More
MSC Fowler 54-101-300-1 0 to 300mm Range, 0.01mm Resolution, Electronic Caliper Stainless Steel with 2-13/32" Stainless Steel Jaws, 0.02mm Accuracy
Catalog No. 501113278
Encompass
Fowler 54-101-300-1 0 to 300mm Range, 0.01mm Resolution, Electronic Caliper Stainless Steel with 2-13/32" Stainless Steel Jaws, 0.02mm Accuracy
Catalog No. 50-111-3278 Supplier MSC Supplier No. 77750701
Fowler 54-101-300-1 0 to 300mm Range, 0.01mm Resolution, Electronic Caliper Stainless Steel with 2-13/32" Stainless Steel Jaws, 0.02mm Accuracy