Learn More
MSC Stanco Temp-Test Bib Overalls, Indura Ultrasoft, 12.4 cal/cm², 5XL, 1 ea
Catalog No. 5000688736
Encompass
These electrical arc protection bib overalls provide dependable comfort and safety in high-risk environments. Constructed from 9 oz. Indura Ultra Soft flame-resistant material, these garments are engineered to meet strict safety standards while maintaining durability through a 5-thread safety stitch design.
  • Meets NFPA 70E and ASTM F1506 specifications
  • ASTM 2178 certified
  • Arc thermal performance value of 12.4 cal/cm2
  • HRC level 2 garment
  • Features velcro front leg enclosures and flame-resistant trim
  • Includes adjustable leg take-ups and elastic shoulder straps for a secure fit
  • Adjustable waist accommodates sizes 34 in. to 54 in.
Catalog No. 50-006-88736 Supplier MSC Supplier No. 93254902
These electrical arc protection bib overalls provide dependable comfort and safety in high-risk environments. Constructed from 9 oz. Indura Ultra Soft flame-resistant material, these garments are engineered to meet strict safety standards while maintaining durability through a 5-thread safety stitch design.
  • Meets NFPA 70E and ASTM F1506 specifications
  • ASTM 2178 certified
  • Arc thermal performance value of 12.4 cal/cm2
  • HRC level 2 garment
  • Features velcro front leg enclosures and flame-resistant trim
  • Includes adjustable leg take-ups and elastic shoulder straps for a secure fit
  • Adjustable waist accommodates sizes 34 in. to 54 in.