Learn More
Ace Glass, Inc. 12/5 ball joint, stainless steel, unthreaded
SDP
Catalog No. 50975293
Encompass
12/5, unthreaded, ball joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.
Catalog No. 50-975-293 Supplier Ace Glass, Inc. Supplier No. 765808
12/5, unthreaded, ball joint fabricated from 18-8 type 303 stainless steel. Fully interchangeable with ACE glass joints and has the same I.D.