Learn More
Chemglass Life Sciences Joint, 12/5, Polished Socket, Clamp Size 12, 7.80-8.20mm Tubing
SDP
Catalog No. CG121P01
Encompass_Preferred
Joint, 12/5, Polished Socket, Clamp Size 12, 7.80-8.20mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.
Catalog No. CG121P01 Supplier Chemglass Life Sciences Supplier No. CG121P01
Joint, 12/5, Polished Socket, Clamp Size 12, 7.80-8.20mm Tubing. For use with CG-122, o-ring ball joint to provide for a flexible high vacuum seal. Polished sealing surface does not abrade o-ring. Note: Not for use with CG-120 ground ball joints.