Learn More
Chemglass Life Sciences 12/5 Socket Joint, 8.0mm OD Tube, Clamp Size = 12
SDP
Catalog No. 501531911
Encompass_Preferred
12/5 Socket Joint, 8.0mm OD Tube, Clamp Size = 12. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.
Catalog No. 50-153-1911 Supplier Chemglass Life Sciences Supplier No. CGB12104
12/5 Socket Joint, 8.0mm OD Tube, Clamp Size = 12. Made to ASTM Specifications E677. See CG-145-B and CG-150 for clamps.