Learn More
Ace Glass, Inc. 12/5 socket joint, stainless steel, 1/8" male NPT
SDP
Catalog No. 50975304
Encompass
12/5 socket joint, fabricated from 18-8 type 303 stainless steel, is fully interchangeable with ACE glass joints, and has same size I.D. Shank is machined to 1/8 male NPT.
Catalog No. 50-975-304 Supplier Ace Glass, Inc. Supplier No. 7658210
12/5 socket joint, fabricated from 18-8 type 303 stainless steel, is fully interchangeable with ACE glass joints, and has same size I.D. Shank is machined to 1/8 male NPT.