Learn More
Integra Biosciences Corp 12.5ML STERILE TIPS 384/CS
SDP
Catalog No. NC1084183
Encompass
GripTips snap onto the tip fitting of the pipetting head, which prevents tips from falling off • An additional quality check ensures that pipette tips are perfectly straight, enabling precise pipetting in multi well plates • Filtered GripTips reliably protect the pipetting head against contaminations caused by aerosols
Catalog No. NC1084183 Supplier Integra Biosciences Corp Supplier No. 6454
GripTips snap onto the tip fitting of the pipetting head, which prevents tips from falling off • An additional quality check ensures that pipette tips are perfectly straight, enabling precise pipetting in multi well plates • Filtered GripTips reliably protect the pipetting head against contaminations caused by aerosols