Learn More
Integra Biosciences Corp 12.5UL,5 XYZ RACKS/384
SDP
Catalog No. NC2239428
Encompass
Precise and consistent tip seal ensures that all tips on MINI 96, VIAFLO 96 and VIAFLO 384 are at an identical height for reliable touch offs and consistent pipetting results. An additional tip straightness inspection process ensures all pipette tips access wells even when dispensing to a 384 well plate.For use with 2, 10, 12.5 and 20 µl INTEGRA pipettes44 mm length for easy access to 1.5 ml tubes
This item is not returnable. View return policy
Catalog No. NC2239428 Supplier Integra Biosciences Corp Supplier No. 6404
Precise and consistent tip seal ensures that all tips on MINI 96, VIAFLO 96 and VIAFLO 384 are at an identical height for reliable touch offs and consistent pipetting results. An additional tip straightness inspection process ensures all pipette tips access wells even when dispensing to a 384 well plate.For use with 2, 10, 12.5 and 20 µl INTEGRA pipettes44 mm length for easy access to 1.5 ml tubes