Learn More
Chemglass Life Sciences Replacement Washers for 12.9/44 PTFE Plug
SDP
Catalog No. CG45708
Encompass_Preferred
Replacement Washers for 12.9/44 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.
Catalog No. CG45708 Supplier Chemglass Life Sciences Supplier No. CG45708
Replacement Washers for 12.9/44 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.