Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

12-Aminolauric Acid 98.0+%, TCI America™
SDP

Catalog No. A0932500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0932500G 500 g
A093225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0932500G Supplier TCI America Supplier No. A0932500G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 693-57-2
Molecular Formula C12H25NO2
Molecular Weight (g/mol) 215.337
MDL Number MFCD00008153
InChI Key PBLZLIFKVPJDCO-UHFFFAOYSA-N
Synonym 12-aminolauric acid, 12-amino-dodecanoic acid, dodecanoic acid, 12-amino, ccris 6171, omega-aminododecanoic acid, 12-aminolauricacid, omega-aminolauric acid, 12-aminododecanic acid, .omega.-aminolauric acid
PubChem CID 69661
IUPAC Name 12-aminododecanoic acid
SMILES C(CCCCCC(=O)O)CCCCCN

Specifications

Melting Point 186°C
Color White
Quantity 500 g
Formula Weight 215.34
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 12-Aminolauric Acid
EINECSNumber (2)-3121
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.