Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 12-azido-dodecanoyl-OSu | 2489524-00-5 | ≥97.0% | 338.40 g/mol | C16H26N4O4 | 10 MG
SDP

Catalog No. 5000286608
Encompass_Preferred

12-Azido-dodecanoyl-OSu is an azide-functionalized N-hydroxysuccinimide (OSu) ester used as a hydrophobic linker for bioconjugation and click-chemistry applications. The reagent introduces a 12-carbon azidododecanoyl group that facilitates azide-alkyne cycloaddition and enables efficient conjugation to primary amines via NHS-ester chemistry. Supplied as a solid with high purity for labeling and linker synthesis.

  • Azide-functionalized OSu ester for click chemistry.
  • Reactive toward primary amines for amide bond formation.
  • Hydrophobic 12-carbon spacer useful for membrane-targeting conjugates.
  • Purity ≥97.0%.
  • Molecular weight 338.40 g/mol; formula C16H26N4O4.
  • Available in small research sizes, including 10 mg.
  • Store sealed, away from moisture; solid at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-86608 Supplier Medchemexpress LLC Supplier No. HY15181810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

12-Azido-dodecanoyl-OSu is an azide-functionalized N-hydroxysuccinimide (OSu) ester used as a hydrophobic linker for bioconjugation and click-chemistry applications. The reagent introduces a 12-carbon azidododecanoyl group that facilitates azide-alkyne cycloaddition and enables efficient conjugation to primary amines via NHS-ester chemistry. Supplied as a solid with high purity for labeling and linker synthesis.

  • Azide-functionalized OSu ester for click chemistry.
  • Reactive toward primary amines for amide bond formation.
  • Hydrophobic 12-carbon spacer useful for membrane-targeting conjugates.
  • Purity ≥97.0%.
  • Molecular weight 338.40 g/mol; formula C16H26N4O4.
  • Available in small research sizes, including 10 mg.
  • Store sealed, away from moisture; solid at 4°C; in solution store at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.