Learn More
Medchemexpress LLC 12-Azido-dodecanoyl-OSu | 2489524-00-5 | C16H26N4O4 | 50 MG
SDP
Catalog No. 5000374860
Encompass_Preferred
12-Azido-dodecanoyl-OSu is a click chemistry reagent containing an azide group, primarily used as a hydrophobic bioconjugation linker.
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have alkyne groups.
  • Can also participate in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.
Catalog No. 50-003-74860 Supplier Medchemexpress LLC Supplier No. HY15181850MG
12-Azido-dodecanoyl-OSu is a click chemistry reagent containing an azide group, primarily used as a hydrophobic bioconjugation linker.
  • Capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that have alkyne groups.
  • Can also participate in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules containing DBCO or BCN groups.