Learn More
FUJIFILM Biosciences 12-Bis bis 35-dit-butylphenyl
Catalog No. 5000450208
Encompass_Preferred
Bulky bidentate phosphine ligand used in transition metal catalysis This is a ligand for Suzuki-Miyaura coupling reaction It is made into an iron complex to be used in the coupling reaction Soluble in acetone slightly soluble in ethanol and water White to nearly white crystalline powder For research use only RUO
Catalog No. 50-004-50208 Supplier FUJIFILM Biosciences Supplier No. 02517083
Bulky bidentate phosphine ligand used in transition metal catalysis This is a ligand for Suzuki-Miyaura coupling reaction It is made into an iron complex to be used in the coupling reaction Soluble in acetone slightly soluble in ethanol and water White to nearly white crystalline powder For research use only RUO