Learn More
Bio Rad Laboratories 12% BIS-TRIS GEL 18W
Catalog No. NC9081975
Encompass
Criterion XT Bis-Tris Gel, 12%, 18-well, 30microliter, 13.3cm x 8.7cm, Solid, White, Odorless, Boiling point: 100degreeC, Flash point: >100degreeC, pH: 6.4 at 20degreeC, Ignition temperature: 400.0degreeC
Catalog No. NC9081975 Supplier Bio Rad Laboratories Supplier No. 3450118
Criterion XT Bis-Tris Gel, 12%, 18-well, 30microliter, 13.3cm x 8.7cm, Solid, White, Odorless, Boiling point: 100degreeC, Flash point: >100degreeC, pH: 6.4 at 20degreeC, Ignition temperature: 400.0degreeC