Learn More
Chemscene CS-0106409,AbaChemscene,(-)-1,2-Bis[(2S,5S)-2,5-diisopropylphospholano]benzene,147253-69-8,100mg
SDP
Supplier:  Chemscene CS0106409100MG
Encompass_Preferred
CS-0106409,AbaChemscene,(-)-1,2-Bis[(2S,5S)-2,5-diisopropylphospholano]benzene,147253-69-8,100mg,Formula:C26H44P2,M. Wt. :418.58,Purity:>98%,Smiles,CC(C)[C@H]1P(C2=CC=CC=C2P3[C@H](C(C)C)CC[C@H]3C(C)C)[C@H](C(C)C)CC1.[(S,S)]
Catalog No. 50-209-3561