Learn More
Accela Chembio Inc MF: C12H25BrO | MW: 265.23 | >95% | Flash Point: 113 celsius | BP: 155 celsius at 760 mmHg.
Catalog No. 50481327
Encompass_Preferred
MF: C12H25BrO | MW: 265.23 | >95% | Flash Point: 113 celsius | BP: 155 celsius at 760 mmHg.
Catalog No. 50-481-327 Supplier Accela Chembio Inc Supplier No. SY01946825G
MF: C12H25BrO | MW: 265.23 | >95% | Flash Point: 113 celsius | BP: 155 celsius at 760 mmHg.