Learn More
Medchemexpress LLC 12-Bromododecanoic a 25g
SDP
Supplier:  Medchemexpress LLC 25GHYW105741
Encompass_Preferred
12-Bromododecanoic acid is a bromo fatty acid 12-Bromododecanoic acid can be used in the synthesis of clickable forms of myristic acid 12-Bromododecanoic acid inhibits HIV replication in CEM-SS T cells (EC50 38 M)[1][2]
Catalog No. 50-002-05453