Learn More
Cayman Chemical 12-Bromododecanoic Acid, Purity: ≥95%, CAS: 73367-80-3, 500 mg
SDP
Catalog No. 502992039
Encompass_Preferred
A halogenated form of lauric acid; has been used in the synthesis of clickable forms of myristic acid and as a model fatty acid ligand to determine the X-ray crystal structure of bovine β-lactoglobulin-ligand complexes; reduces virion DNA in the culture supernatant of primary hepatocytes isolated from a duckling model of HBV infection at 10 µg/ml and inhibits HIV replication in CEM-SS T cells (EC50 = 38 µM)
Catalog No. 50-299-2039 Supplier Cayman Chemical Supplier No. 30465500 MG
A halogenated form of lauric acid; has been used in the synthesis of clickable forms of myristic acid and as a model fatty acid ligand to determine the X-ray crystal structure of bovine β-lactoglobulin-ligand complexes; reduces virion DNA in the culture supernatant of primary hepatocytes isolated from a duckling model of HBV infection at 10 µg/ml and inhibits HIV replication in CEM-SS T cells (EC50 = 38 µM)