Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ 12 Cal ArcGuard™ UltraSoft Arc Flash Leggings
SDP

Catalog No. 170012049 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Large
X-Large
2X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-2049 2X-Large
17-001-2047 Large
17-001-2048 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 17-001-2049 Supplier National Safety Apparel™ Supplier No. L05UP2X
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ultrasoft flame resistant leggings are comfortable and typically paired with an arc rated long coat. Compliant to NFPA 70E with an arc rating of 12 cal/cm² and an CAT level 2.

  • 9 oz. UltraSoft™
  • Adjustable FR hook and loop closure
  • Single layer legging with foot flap
  • For use with 12 cal/cm2 FR long coat

Specifications

Size 2X-Large
Gender Unisex
Material Cotton, Nylon
Color Navy
Ankle Style Open
Seam Type Stitched
Disposable Reusable
Arc Flash Rating 12 cal/cm²
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462
Sterility Non-sterile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.