Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ 12 Cal Hover™ Series Faceshield with Hard Hat
SDP

Catalog No. 17355209 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Arc Flash Rating:
12 cal/cm²
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating
17-355-209 12 cal/cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-355-209 Supplier National Safety Apparel™ Supplier No. H12HTHAT
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Slotted adapter provides enhanced visibility and flexibility.

  • Premium Hover™ series bracket for weight compensation over hard hat
  • Hard hat is included
  • Highly transparent arc flash faceshield for greater visibility
  • Proven color recognition
  • Low profile chin cup
  • Anti-fog and scratch resistant coating
  • Adjustable settings allow the faceshield to be secured in various positions
  • Lightweight design

Specifications

Product Type Arc Flash Face Shield
Size Universal
Material Plastic
Color White, Clear
Disposable Reusable
Arc Flash Rating 12 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F2178, CSA Z462, ANSI/ISEA Z87.1, OSHA 1910.269
Quantity 1 Ea.
Includes Hard Hat
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Style Faceshield
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.