Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ 12 cal Arc Flash Faceshield Kit
SDP

Catalog No. 17355207 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Arc Flash Rating:
12 cal/cm²
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Arc Flash Rating
17-355-207 12 cal/cm²
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 17-355-207 Supplier National Safety Apparel™ Supplier No. KITHP12PV
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Arc flash faceshield kit includes a clear, low profile chin cup that increases field of vision and offers protection when looking down.

  • Premium Hover series bracket for weight compensation over hard hat
  • Highly transparent arc flash faceshield for greater visibility
  • Proven color recognition
  • Low profile chin cup
  • Anti-fog and scratch resistant coating
  • Adjustable settings allow the faceshield to be secured in various positions
  • Lightweight design

Specifications

Product Type Arc Flash Head Protection Kit
Size Universal
Material Plastic, UltraSoft
Color Clear
Disposable Reusable
Arc Flash Rating 12 cal/cm²
Certifications/Compliance NFPA 70E, ASTM F1506, ASTM F2178, CSA Z462, ANSI/ISEA Z87.1, OSHA 1910.269
Quantity 1 Ea.
Includes 12 cal lift front faceshield, balaclava, and bag
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Style Faceshield Kit
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.