Learn More
National Safety Apparel™ 12 Cal UltraSoft Arc Flash Coat
SDP
Catalog No. 17001229 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-229 Small
17-001-230 Medium
17-001-231 Large
17-001-232 2X-Large
17-001-233 3X-Large
17-001-234 X-Large
17-001-235 4X-Large
17-001-236 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-229 Supplier National Safety Apparel™ Supplier No. C04UP03SM32
Add to Cart
Add to Cart
For enhanced protection Westex™ UltraSoft is the ideal choice for superior comfort, improved safety, and above average durability.
  • Arc Rating: 12 cal/cm²
  • CAT 2
  • Fabric/Material: 9 oz. Westex™ UltraSoft™
  • 32 in. coat length
  • Garments embroidered with arc rating
  • FR hook and loop front closure
  • FR knit wrists for comfortable, secure fit
  • FR fabric will not melt, drip, or ignite
  • Generous fit for ease of movement
  • Stand up collar for extra coverage
  • Available in a convenient kit
  • Made in the USA
Specifications
Closure Type Hook and Loop
Color Navy
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size Small
Resistance Flame
Seam Type Stitched
Arc Flash Rating 8 cal/cm²
Certifications/Compliance CAT 2, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269