Learn More
National Safety Apparel™ 12 Cal UltraSoft Arc Flash Coat
SDP
Catalog No. 17001236 Shop All National Safety Apparel Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-236 5X-Large
17-001-229 Small
17-001-230 Medium
17-001-231 Large
17-001-232 2X-Large
17-001-233 3X-Large
17-001-234 X-Large
17-001-235 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-236 Supplier National Safety Apparel™ Supplier No. C04UP035X32
For enhanced protection Westex™ UltraSoft is the ideal choice for superior comfort, improved safety, and above average durability.
  • Arc Rating: 12 cal/cm²
  • CAT 2
  • Fabric/Material: 9 oz. Westex™ UltraSoft™
  • 32 in. coat length
  • Garments embroidered with arc rating
  • FR hook and loop front closure
  • FR knit wrists for comfortable, secure fit
  • FR fabric will not melt, drip, or ignite
  • Generous fit for ease of movement
  • Stand up collar for extra coverage
  • Available in a convenient kit
  • Made in the USA
Specifications
Closure Type Hook and Loop
Color Navy
Disposable Reusable
Gender Unisex
Material Cotton, Nylon
Neck Style Collared
Size 5X-Large
Resistance Flame
Seam Type Stitched
Arc Flash Rating 8 cal/cm²
Certifications/Compliance CAT 2, NFPA 70 E, CSA Z462, ASTM F1506, OSHA 1910.269