Learn More
National Safety Apparel™ 12 Cal UltraSoft Bib Overalls
SDP
Catalog No. 17001341 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-001-341 Small
17-001-342 Medium
17-001-343 Large
17-001-344 X-Large
17-001-345 2X-Large
17-001-346 3X-Large
17-001-347 4X-Large
17-001-348 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 17-001-341 Supplier National Safety Apparel™ Supplier No. C45UPSM32
Add to Cart
Add to Cart
Overalls feature durable water-repellent finish. Compliant with inherent FR protection.
  • Arc Rating: 12 cal/cm²
  • CAT 2
  • Fabric/Material: 9 oz. Westex™ UltraSoft™
  • 12 cal FR UltraSoft 32 in. Bib Overalls Quick disconnect and easy adjustable shoulder straps for quick entry
  • FR hook and loop fly closure
  • FR hook and loop calf closure for easy boot entry
  • FR fabric will not melt, drip, or ignite
  • Rear pockets
  • Expandable leg opening to fit over boots
  • Convenient side pass-through pockets for access to pants pockets
  • Garments embroidered with arc rating
  • Available in a convenient kit
  • Made in the USA.
Specifications
Closure Type Buckle
Color Navy
Material Cotton, Nylon
Seam Type Stitched
Size Small
Suspender Adjustable
Ankle Style Open
Arc Flash Rating 12 cal/cm²
Waist Style Standard
Resistance Flame
Certifications/Compliance CAT 2, ASTM F1506, OSHA 1910.269, NFPA 70 E, CSA Z462
Disposable Reusable