Learn More
Electron Microscopy Sciences Spot Plate Polypropylene with 12 Cavities
SDP
Catalog No. 50949551
Encompass_Preferred
Very similar to the white porcelain plate, this PP plate comprises 12 cavities of approximately 1ml capacity and is economically priced. This plate is very high quality, unbreakable as well as autoclavable.
Catalog No. 50-949-551 Supplier Electron Microscopy Sciences Supplier No. 7157210
Very similar to the white porcelain plate, this PP plate comprises 12 cavities of approximately 1ml capacity and is economically priced. This plate is very high quality, unbreakable as well as autoclavable.