Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 12 Channel, 12 Well, PCR, 19 Gauge, SS Tubes, 7mm Long, 9mm Spacing, Polycarbonate
Catalog No. NC1555819
Encompass
ASPIRATION MANIFOLD, 12 Channel, 12 Well, PCR Plate, 19 Gauge, Stainless Steel Tubes, 7mm Long, 9mm Center Spacing, Polycarbonate, Female Luer Lok
Catalog No. NC1555819 Supplier V&P Scientific Inc Supplier No. VP 187PC1S
ASPIRATION MANIFOLD, 12 Channel, 12 Well, PCR Plate, 19 Gauge, Stainless Steel Tubes, 7mm Long, 9mm Center Spacing, Polycarbonate, Female Luer Lok