Learn More
USA Scientific Inc 12-well V-bottom reservoir, sterile
Catalog No. NC9632753
Encompass
Twelve well v-bottom reservoir with lid, individually wrapped, 20/pack, sterile. Each v-shaped well holds up to 3 mL.
Catalog No. NC9632753 Supplier USA Scientific Inc Supplier No. 38233512
Twelve well v-bottom reservoir with lid, individually wrapped, 20/pack, sterile. Each v-shaped well holds up to 3 mL.