Learn More
V&P Scientific Inc ASPIRATION WAND, 12 Channel, 96 Well, Standard, 21 Gauge, SS Tubes, 11mm Long, 9mm Spacing, Polycarbonate
Catalog No. NC9987619
Encompass
ASPIRATION WAND, 12 Channel, 96 Well, Standard Plate, 21 Gauge, Stainless Steel Tubes, 11mm Long, 9mm Center Spacing, Polycarbonate, Female Luer Lok
Catalog No. NC9987619 Supplier V&P Scientific Inc Supplier No. VP 185L1
ASPIRATION WAND, 12 Channel, 96 Well, Standard Plate, 21 Gauge, Stainless Steel Tubes, 11mm Long, 9mm Center Spacing, Polycarbonate, Female Luer Lok