Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well, 19 Gauge, SS Tubes, 30mm Long, 9mm Spacing, Polypropylene
Catalog No. NC9012061
Encompass
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 30mm Long, 9mm Center Spacing, Polypropylene
Catalog No. NC9012061 Supplier V&P Scientific Inc Supplier No. VP 187BP
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 30mm Long, 9mm Center Spacing, Polypropylene