Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well, 19 Gauge, SS Tubes, 44mm Long, 9mm Spacing, Polycarbonate
Catalog No. NC0489420
Encompass
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 44mm Long, 9mm Center Spacing, Polycarbonate, Female Luer Lok
Catalog No. NC0489420 Supplier V&P Scientific Inc Supplier No. VP 187B
Add to Cart
Add to Cart
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 44mm Long, 9mm Center Spacing, Polycarbonate, Female Luer Lok