Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well, 19 Gauge, SS Tubes, 60mm Long, 9mm Spacing, Polypropylene
Catalog No. NC0407305
Encompass
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 60mm Long, 9mm Center Spacing, Polypropylene, Female Luer Lok
Catalog No. NC0407305 Supplier V&P Scientific Inc Supplier No. VP 187BP60
ASPIRATION MANIFOLD, 12 Channel, 12 Well, Deep Well Plate, 19 Gauge, Stainless Steel Tubes, 60mm Long, 9mm Center Spacing, Polypropylene, Female Luer Lok