Learn More
ANALYTIK JENA US LLC Analytik Jena 12-Channel Adapter, Liquid Handling, Compatible With CyBio FeliX Head R 96, Adapter
Catalog No. NC3734417
Encompass
The Analytik Jena 12-Channel Adapter enhances liquid handling efficiency by enabling precise 12- to 1-channel transfers within the CyBio FeliX Head R 96 system. It supports sample preparation, compound handling, and assay implementation across 96- and 384-well microplates, making it a versatile tool for various laboratory applications. This adapter stands out for its modular design, ensuring high precision while minimizing reagent use, contributing to cost-effective scientific workflows.
Catalog No. NC3734417 Supplier ANALYTIK JENA US LLC Supplier No. OL331714340
Add to Cart
Add to Cart
Request a Quote
The Analytik Jena 12-Channel Adapter enhances liquid handling efficiency by enabling precise 12- to 1-channel transfers within the CyBio FeliX Head R 96 system. It supports sample preparation, compound handling, and assay implementation across 96- and 384-well microplates, making it a versatile tool for various laboratory applications. This adapter stands out for its modular design, ensuring high precision while minimizing reagent use, contributing to cost-effective scientific workflows.