Learn More
Waters Corp Andrew Alliance Pipette, 12-channel, 10-300 μL
Catalog No. 502269337 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Andrew Alliance Bluetooth Electronic 12-channel Pipettes provide accurate and precise pipetting for use with Pipette+ guided pipetting system or manual pipetting. Based on the Sartorius Picus Pipette, the ergonomic and lightweight design helps protect users from repetitive strain injuries by ensuring an optimal working posture and comfortable pipetting. Fully electronic operation reduces variability between users. Bluetooth capability allows two-way communication with pipettes, when used with Pipette+, and OneLab enabling remote programming of pipettes with the added advantage of pipetting traceability.
  • Intuitive user interface.
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts).
  • Optiload for perfect tip sealing.
  • Safe-cone filters in all models >10 μL to prevent contamination.
This item is not returnable. View return policy
Catalog No. 50-226-9337 Supplier Waters Corp Supplier No. 186010104
Andrew Alliance Bluetooth Electronic 12-channel Pipettes provide accurate and precise pipetting for use with Pipette+ guided pipetting system or manual pipetting. Based on the Sartorius Picus Pipette, the ergonomic and lightweight design helps protect users from repetitive strain injuries by ensuring an optimal working posture and comfortable pipetting. Fully electronic operation reduces variability between users. Bluetooth capability allows two-way communication with pipettes, when used with Pipette+, and OneLab enabling remote programming of pipettes with the added advantage of pipetting traceability.
  • Intuitive user interface.
  • Autoclavable lower parts (except for 1200 μL multi-channel lower parts).
  • Optiload for perfect tip sealing.
  • Safe-cone filters in all models >10 μL to prevent contamination.