Learn More
Waters Corp Andrew Alliance Pipette, 12-channel, 0.2-10 μL
Catalog No. 502269335 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Andrew Alliance Bluetooth Electronic 12-channel Pipettes offer accurate and precise pipetting, designed for use with the Pipette+ guided pipetting system or for manual operation. Based on the Sartorius Picus Pipette, its ergonomic and lightweight design aims to prevent repetitive strain injuries by promoting an optimal working posture and comfortable pipetting. The fully electronic operation helps reduce variability between users. Its Bluetooth capability enables two-way communication with pipettes when used with Pipette+ and OneLab, allowing remote programming of pipettes and providing pipetting traceability.
  • Intuitive user interface.
  • Autoclavable lower parts (excluding 1200 μL multi-channel lower parts).
  • Optiload for perfect tip sealing.
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing, and more.
This item is not returnable. View return policy
Catalog No. 50-226-9335 Supplier Waters Corp Supplier No. 186010102
Andrew Alliance Bluetooth Electronic 12-channel Pipettes offer accurate and precise pipetting, designed for use with the Pipette+ guided pipetting system or for manual operation. Based on the Sartorius Picus Pipette, its ergonomic and lightweight design aims to prevent repetitive strain injuries by promoting an optimal working posture and comfortable pipetting. The fully electronic operation helps reduce variability between users. Its Bluetooth capability enables two-way communication with pipettes when used with Pipette+ and OneLab, allowing remote programming of pipettes and providing pipetting traceability.
  • Intuitive user interface.
  • Autoclavable lower parts (excluding 1200 μL multi-channel lower parts).
  • Optiload for perfect tip sealing.
  • Optimized pipetting modes for multi-dispensing, multi-aspiration, reverse pipetting, mixing, and more.