Learn More
V&P Scientific Inc ASPIRATION WAND, 12 Channel, 96 Well, Standard, 20 Gauge, SS Tubes, 11mm Long, 9mm Spacing, Polycarbonate
Catalog No. NC1591222
Encompass
ASPIRATION WAND, 12 Channel, 96 Well, Standard Plate, 20 Gauge, Stainless Steel Tubes, 11mm Long, 9mm Center Spacing, Polycarbonate. Female Luer Lok
Catalog No. NC1591222 Supplier V&P Scientific Inc Supplier No. VP 185LA
ASPIRATION WAND, 12 Channel, 96 Well, Standard Plate, 20 Gauge, Stainless Steel Tubes, 11mm Long, 9mm Center Spacing, Polycarbonate. Female Luer Lok